Furo[3,2-b]pyridine-6-carbaldehyde
Supplier: Matrix Scientific
035776-100MG
035776-500MG
101840-032EA
364.62
USD
101840-032
101840-034
Furo[3,2-b]pyridine-6-carbaldehyde
Furo[3,2-b]pyridine-6-carbaldehyde
Hazard: Irritant
Formula:
C₈H₅NO₂ |
MDL Number:
MFCD11857749 CAS Number: 227939-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...