(6-Bromofuro[3,2-b]pyridin-2-yl)methanol

Supplier: Matrix Scientific

035771-100MG 035771-500MG
101840-012EA 273.85 USD
101840-012 101840-014
(6-Bromofuro[3,2-b]pyridin-2-yl)methanol
(6-Bromofuro[3,2-b]pyridin-2-yl)methanol

Hazard: Irritant

Formula: C₈H₆BrNO₂
MDL Number: MFCD11857745
CAS Number: 1131335-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR