3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

035770-100MG 035770-500MG
101840-008EA 364.62 USD
101840-008 101840-010
3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
3-Iodo-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₄F₃IN₂
MW: 312.03 g/mol
MDL Number: MFCD11857744
CAS Number: 1142192-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR