5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Supplier: Matrix Scientific

035768-100MG 035768-500MG
101840-000EA 364.62 USD
101840-000 101840-002
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
5-(Trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₅F₃N₂
MW: 186.1300 g/mol
MDL Number: MFCD06659680
CAS Number: 1036027-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR