2-Chloro-3-(dimethoxymethyl)isonicotinaldehyde

Supplier: Matrix Scientific

035765-100MG 035765-500MG
101839-988EA 364.62 USD
101839-988 101839-990
2-Chloro-3-(dimethoxymethyl)isonicotinaldehyde
2-Chloro-3-(dimethoxymethyl)isonicotinaldehyde

Hazard: Irritant

Formula: C₉H₁₀ClNO₃
MDL Number: MFCD12026769
CAS Number: 1142192-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR