tert-Butyl (6-bromo-2-chloropyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

035762-100MG 035762-500MG
101839-976EA 364.62 USD
101839-976 101839-978
tert-Butyl (6-bromo-2-chloropyridin-3-yl)methylcarbamate
tert-Butyl (6-bromo-2-chloropyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₁H₁₄BrClN₂O₂
MW: 321.61 g/mol
MDL Number: MFCD12026767
CAS Number: 1142192-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR