N-(4-Iodo-2-methoxypyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035757-100MG 035757-500MG
101839-956EA 364.62 USD
101839-956 101839-958
N-(4-Iodo-2-methoxypyridin-3-yl)pivalamide
N-(4-Iodo-2-methoxypyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₅IN₂O₂
MDL Number: MFCD12026762
CAS Number: 131653-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR