tert-Butyl-2-iodo-6-((trimethylsilyl)ethynyl)pyridin-3-yl carbonate

Supplier: Matrix Scientific

035755-100MG 035755-500MG
101839-948EA 364.62 USD
101839-948 101839-950
tert-Butyl-2-iodo-6-((trimethylsilyl)ethynyl)pyridin-3-yl carbonate
tert-Butyl-2-iodo-6-((trimethylsilyl)ethynyl)pyridin-3-yl carbonate

Hazard: Irritant

Formula: C₁₅H₂₀INO₃Si
MW: 417.32 g/mol
MDL Number: MFCD12026760
CAS Number: 1142192-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR