N-(3-Hydroxy-2-iodopyridin-4-yl)pivalamide

Supplier: Matrix Scientific

035754-100MG 035754-500MG
101839-944EA 364.62 USD
101839-944 101839-946
N-(3-Hydroxy-2-iodopyridin-4-yl)pivalamide
N-(3-Hydroxy-2-iodopyridin-4-yl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₃IN₂O₂
MDL Number: MFCD12026759
CAS Number: 1142192-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR