(3R,4R)-1-tert-Butyl-3-methyl-4-(6-chloro-5-pivalamidopyridin-2-yl)pyrrolidine-1,3-dicarboxylate

Supplier: Matrix Scientific

035753-100MG 035753-500MG
101839-940EA 364.62 USD
101839-940 101839-942
(3R,4R)-1-tert-Butyl-3-methyl-4-(6-chloro-5-pivalamidopyridin-2-yl)pyrrolidine-1,3-dicarboxylate
(3R,4R)-1-tert-Butyl-3-methyl-4-(6-chloro-5-pivalamidopyridin-2-yl)pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₂₁H₃₀ClN₃O₅
MDL Number: MFCD12026758
CAS Number: 1217607-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR