tert-Butyl-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate

Supplier: Matrix Scientific

035751-100MG 035751-500MG
101839-932EA 364.62 USD
101839-932 101839-934
tert-Butyl-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate
tert-Butyl-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MW: 249.27 g/mol
MDL Number: MFCD12026756
CAS Number: 1142192-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR