N-(2-Bromo-5-formylpyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035750-100MG 035750-500MG
101839-928EA 364.62 USD
101839-928 101839-930
N-(2-Bromo-5-formylpyridin-3-yl)pivalamide
N-(2-Bromo-5-formylpyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₃BrN₂O₂
MDL Number: MFCD12026755
CAS Number: 1142192-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR