5-(Trifluoromethyl)-3-((trimethylsilyl)ethynyl)pyridin-2-amine

Supplier: Matrix Scientific

035748-100MG 035748-500MG
101839-920EA 364.62 USD
101839-920 101839-922
5-(Trifluoromethyl)-3-((trimethylsilyl)ethynyl)pyridin-2-amine
5-(Trifluoromethyl)-3-((trimethylsilyl)ethynyl)pyridin-2-amine

Hazard: Irritant

Formula: C₁₁H₁₃F₃N₂Si
MW: 258.32 g/mol
MDL Number: MFCD12026753
CAS Number: 1036027-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR