2-Chloro-3-(dimethoxymethyl)-4-iodopyridine
Supplier: Matrix Scientific
035746-100MG
035746-500MG
101839-912EA
364.62
USD
101839-912
101839-914
2-Chloro-3-(dimethoxymethyl)-4-iodopyridine
2-Chloro-3-(dimethoxymethyl)-4-iodopyridine
Hazard: Irritant
Formula:
C₈H₉ClINO₂ |
MDL Number:
MFCD12026751 CAS Number: 1142192-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...