tert-Butyl-2,5-dibromopyridin-3-yl carbonate

Supplier: Matrix Scientific

035745-100MG 035745-500MG
101839-908EA 364.62 USD
101839-908 101839-910
tert-Butyl-2,5-dibromopyridin-3-yl carbonate
tert-Butyl-2,5-dibromopyridin-3-yl carbonate

Hazard: Irritant

Formula: C₁₀H₁₁Br₂NO₃
MW: 353.02 g/mol
MDL Number: MFCD11857742
CAS Number: 1142192-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR