6-Bromo-2-chloronicotinaldehyde oxime

Supplier: Matrix Scientific

035744-100MG 035744-500MG
101839-904EA 364.62 USD
101839-904 101839-906
6-Bromo-2-chloronicotinaldehyde oxime
6-Bromo-2-chloronicotinaldehyde oxime

Hazard: Irritant

Formula: C₆H₄BrClN₂O
MDL Number: MFCD12026750
CAS Number: 1142192-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR