2-Chloro-6-methoxynicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-methoxypyridine-3-carboxylic acid

035742-1G 035742-5G
101839-896EA 355.38 USD
101839-896 101839-898
2-Chloro-6-methoxynicotinic acid
2-Chloro-6-methoxynicotinic acid

Hazard: Irritant

Formula: C₇H₆NO₃Cl
Melting Pt: 230°C
MDL Number: MFCD08236845
CAS Number: 503000-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR