N-(2-Chloro-6-hydroxypyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035739-100MG 035739-500MG
101839-884EA 364.62 USD
101839-884 101839-886
N-(2-Chloro-6-hydroxypyridin-3-yl)pivalamide
N-(2-Chloro-6-hydroxypyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD11857738
CAS Number: 1142192-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR