Methyl-3-cyano-5-methoxyisonicotinate

Supplier: Matrix Scientific

035738-100MG 035738-500MG
101839-880EA 364.62 USD
101839-880 101839-882
Methyl-3-cyano-5-methoxyisonicotinate
Methyl-3-cyano-5-methoxyisonicotinate

Hazard: Irritant

Formula: C₉H₈N₂O₃
MDL Number: MFCD12026749
CAS Number: 1142192-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR