2,3-Dichloro-4-iodopyridine ≥95%
Supplier: Matrix Scientific
035736-1G
035736-5G
101839-872EA
46.15
USD
101839-872
101839-874
2,3-Dichloro-4-iodopyridine ≥95%
2,3-Dichloro-4-iodopyridine
Hazard: Irritant
Formula:
C₅H₂Cl₂IN MW: 273.8900 g/mol |
MDL Number:
MFCD11857737 CAS Number: 889865-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...