6-Bromo-2-chloro-N-methoxy-N-methylnicotinamide

Supplier: Matrix Scientific

035734-100MG 035734-500MG
101839-864EA 364.62 USD
101839-864 101839-866
6-Bromo-2-chloro-N-methoxy-N-methylnicotinamide
6-Bromo-2-chloro-N-methoxy-N-methylnicotinamide

Hazard: Irritant

Formula: C₈H₈BrClN₂O₂
MDL Number: MFCD11857735
CAS Number: 1142192-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR