tert-Butyl-6-chloro-5-pivalamidopyridin-2-ylcarbamate

Supplier: Matrix Scientific

035733-100MG 035733-500MG
101839-860EA 364.62 USD
101839-860 101839-862
tert-Butyl-6-chloro-5-pivalamidopyridin-2-ylcarbamate
tert-Butyl-6-chloro-5-pivalamidopyridin-2-ylcarbamate

Hazard: Irritant

Formula: C₁₅H₂₂ClN₃O₃
MW: 327.81 g/mol
MDL Number: MFCD11857734
CAS Number: 1142192-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR