tert-Butyl (5,6-dimethoxypyridin-2-yl)methylcarbamate

Supplier: Matrix Scientific

035732-100MG 035732-500MG
101839-856EA 364.62 USD
101839-856 101839-858
tert-Butyl (5,6-dimethoxypyridin-2-yl)methylcarbamate
tert-Butyl (5,6-dimethoxypyridin-2-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₄
MDL Number: MFCD11857733
CAS Number: 1142192-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR