N-(2-Chloro-6-(3-hydroxyprop-1-ynyl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035730-100MG 035730-500MG
101839-848EA 364.62 USD
101839-848 101839-850
N-(2-Chloro-6-(3-hydroxyprop-1-ynyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-(3-hydroxyprop-1-ynyl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂O₂
MW: 266.73 g/mol
MDL Number: MFCD11857731
CAS Number: 1142192-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR