Methyl-6-bromo-2-chloronicotinate

Supplier: Matrix Scientific

Synonyms: Methyl 6-bromo-2-chloronicotinate

035729-100MG 035729-500MG
101839-844EA 152.31 USD
101839-844 101839-846
Methyl-6-bromo-2-chloronicotinate
Methyl-6-bromo-2-chloronicotinate

Hazard: Irritant

Formula: C₇H₅BrClNO₂
MW: 250.49 g/mol
MDL Number: MFCD11857730
CAS Number: 1142192-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR