6-Bromo-2-chloropyridin-3-ol

Supplier: Matrix Scientific

035727-100MG 035727-500MG
101839-836EA 72.31 USD
101839-836 101839-838
6-Bromo-2-chloropyridin-3-ol
6-Bromo-2-chloropyridin-3-ol

Hazard: Irritant

Formula: C₅H₃BrClNO
MDL Number: MFCD11857728
CAS Number: 52764-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR