tert-Butyl (5-methoxypyridine-3,4-diyl)bis(methylene)dicarbamate
Supplier: Matrix Scientific
035726-100MG
035726-500MG
101839-832EA
364.62
USD
101839-832
101839-834
tert-Butyl (5-methoxypyridine-3,4-diyl)bis(methylene)dicarbamate
tert-Butyl (5-methoxypyridine-3,4-diyl)bis(methylene)dicarbamate
TCA listed: T
Hazard: Irritant
Formula:
C₁₈H₂₉N₃O₅ MW: 367.45 g/mol |
MDL Number:
MFCD11857727 CAS Number: 1142191-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...