N-(2-Chloro-6-((trimethylsilyl)ethynyl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035725-100MG 035725-500MG
101839-828EA 364.62 USD
101839-828 101839-830
N-(2-Chloro-6-((trimethylsilyl)ethynyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-((trimethylsilyl)ethynyl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂OSi
MDL Number: MFCD11857726
CAS Number: 1142191-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR