tert-Butyl (5-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

035724-100MG 035724-500MG
101839-824EA 364.62 USD
101839-824 101839-826
tert-Butyl (5-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)methylcarbamate
tert-Butyl (5-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₈H₂₉BN₂O₅
MDL Number: MFCD11857725
CAS Number: 1247726-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR