(Rac-trans)-1-tert-Butyl-3-methyl-4-(5,6-dimethoxypyridin-2-yl)pyrrolidine-1,3-dicarboxylate

Supplier: Matrix Scientific

035721-100MG 035721-500MG
101839-812EA 364.62 USD
101839-812 101839-814
(Rac-trans)-1-tert-Butyl-3-methyl-4-(5,6-dimethoxypyridin-2-yl)pyrrolidine-1,3-dicarboxylate
(Rac-trans)-1-tert-Butyl-3-methyl-4-(5,6-dimethoxypyridin-2-yl)pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₆
MDL Number: MFCD12026747
CAS Number: 1217674-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR