N-(2-Chloro-6-cyanopyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035719-100MG 035719-500MG
101839-804EA 364.62 USD
101839-804 101839-806
N-(2-Chloro-6-cyanopyridin-3-yl)pivalamide
N-(2-Chloro-6-cyanopyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₂ClN₃O
MDL Number: MFCD11857721
CAS Number: 1142191-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR