1-tert-Butyl-3-methyl-4-(2-chloro-4-iodopyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Supplier: Matrix Scientific

035715-100MG 035715-500MG
101839-788EA 364.62 USD
101839-788 101839-790
1-tert-Butyl-3-methyl-4-(2-chloro-4-iodopyridin-3-yl)pyrrolidine-1,3-dicarboxylate
1-tert-Butyl-3-methyl-4-(2-chloro-4-iodopyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀ClIN₂O₄
MDL Number: MFCD11857717
CAS Number: 1257850-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR