6-Bromo-2-chloro-3-(trimethylsilyl)pyridine

Supplier: Matrix Scientific

035713-100MG 035713-500MG
101839-780EA 364.62 USD
101839-780 101839-782
6-Bromo-2-chloro-3-(trimethylsilyl)pyridine
6-Bromo-2-chloro-3-(trimethylsilyl)pyridine

Hazard: Irritant

Formula: C₈H₁₁BrClNSi
MDL Number: MFCD11857715
CAS Number: 1142191-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR