6-Chloro-5-pivalamidopicolinic acid

Supplier: Matrix Scientific

035712-100MG 035712-500MG
101839-776EA 698.46 USD
101839-776 101839-778
6-Chloro-5-pivalamidopicolinic acid
6-Chloro-5-pivalamidopicolinic acid

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃
MDL Number: MFCD11857714
CAS Number: 1142191-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR