tert-Butyl (4,5-dimethoxypyridin-3-yl)methylcarbamate

Supplier: Matrix Scientific

035710-100MG 035710-500MG
101839-768EA 364.62 USD
101839-768 101839-770
tert-Butyl (4,5-dimethoxypyridin-3-yl)methylcarbamate
tert-Butyl (4,5-dimethoxypyridin-3-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₄
MW: 268.32 g/mol
MDL Number: MFCD11857712
CAS Number: 1142191-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR