6-Bromo-2,3-dichloropyridine
Supplier: Matrix Scientific
035709-100MG
035709-500MG
101839-764EA
172.31
USD
101839-764
101839-766
6-Bromo-2,3-dichloropyridine
6-Bromo-2,3-dichloropyridine
Hazard: Irritant
Formula:
C₅H₂BrCl₂N |
CAS Number:
1033820-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...