6-Bromo-2-chloronicotinaldehyde

Supplier: Matrix Scientific

035707-100MG 035707-500MG
101839-756EA 278.46 USD
101839-756 101839-758
6-Bromo-2-chloronicotinaldehyde
6-Bromo-2-chloronicotinaldehyde

Hazard: Irritant

Formula: C₆H₃BrClNO
MDL Number: MFCD11857709
CAS Number: 1125410-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR