N-(2-Chloro-6-formylpyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035706-100MG 035706-500MG
101839-752EA 364.62 USD
101839-752 101839-754
N-(2-Chloro-6-formylpyridin-3-yl)pivalamide
N-(2-Chloro-6-formylpyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MDL Number: MFCD11857708
CAS Number: 1142191-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR