N-(2-Chloro-6-(trimethylsilyl)pyridin-3-yl)pivalamide

Supplier: Matrix Scientific

035705-100MG 035705-500MG
101839-748EA 364.62 USD
101839-748 101839-750
N-(2-Chloro-6-(trimethylsilyl)pyridin-3-yl)pivalamide
N-(2-Chloro-6-(trimethylsilyl)pyridin-3-yl)pivalamide

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂OSi
MW: 284.86 g/mol
MDL Number: MFCD11857707
CAS Number: 1142191-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR