(2-Chloro-4-iodo-3-pyridinyl)methanol

Supplier: Matrix Scientific

035702-100MG 035702-500MG
101839-736EA 364.62 USD
101839-736 101839-738
(2-Chloro-4-iodo-3-pyridinyl)methanol
(2-Chloro-4-iodo-3-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₅ClINO
MW: 269.47 g/mol
MDL Number: MFCD03095296
CAS Number: 884494-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR