2,6-Diiodopyridin-3-ol

Supplier: Matrix Scientific

035699-100MG 035699-500MG
101839-724EA 364.62 USD
101839-724 101839-726
2,6-Diiodopyridin-3-ol
2,6-Diiodopyridin-3-ol

Hazard: Irritant

Formula: C₅H₃I₂NO
MW: 346.89 g/mol
MDL Number: MFCD03265756
CAS Number: 14764-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR