4-Methoxy-3-((trimethylsilyl)ethynyl)pyridin-2-amine

Supplier: Matrix Scientific

035698-100MG 035698-500MG
101839-720EA 364.62 USD
101839-720 101839-722
4-Methoxy-3-((trimethylsilyl)ethynyl)pyridin-2-amine
4-Methoxy-3-((trimethylsilyl)ethynyl)pyridin-2-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₂OSi
MDL Number: MFCD11857672
CAS Number: 1142191-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR