6-(Dimethoxymethyl)-2,3-dimethoxypyridine

Supplier: Matrix Scientific

035697-100MG 035697-500MG
101839-716EA 364.62 USD
101839-716 101839-718
6-(Dimethoxymethyl)-2,3-dimethoxypyridine
6-(Dimethoxymethyl)-2,3-dimethoxypyridine

Hazard: Irritant

Formula: C₁₀H₁₅NO₄
MDL Number: MFCD11857666
CAS Number: 1142191-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR