2,5-Dibromo-3-methoxypyridine

Supplier: Matrix Scientific

035696-100MG 035696-500MG
101839-712EA 160 USD
101839-712 101839-714
2,5-Dibromo-3-methoxypyridine
2,5-Dibromo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₅Br₂NO
CAS Number: 1142191-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR