2,5-Dibromopyridin-3-ol

Supplier: Matrix Scientific

035695-100MG 035695-500MG
101839-708EA 364.62 USD
101839-708 101839-710
2,5-Dibromopyridin-3-ol
2,5-Dibromopyridin-3-ol

Hazard: Irritant

Formula: C₅H₃Br₂NO
MDL Number: MFCD09959722
CAS Number: 857429-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR