2-Bromo-5-iodopyridin-3-ol

Supplier: Matrix Scientific

035691-100MG 035691-500MG
101839-692EA 364.62 USD
101839-692 101839-694
2-Bromo-5-iodopyridin-3-ol
2-Bromo-5-iodopyridin-3-ol

Hazard: Irritant

Formula: C₅H₃BrINO
MDL Number: MFCD12026744
CAS Number: 1142191-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR