1-Bromo-4-(difluoromethyl)benzene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromobenzal fluoride

035680-1G 035680-5G 035680-25G
101839-656EA 161.54 USD
101839-656 101839-658 101839-660
1-Bromo-4-(difluoromethyl)benzene ≥98%
1-Bromo-4-(difluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 100°C@53mm
MDL Number: MFCD06657964
CAS Number: 51776-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR