1-Bromo-2-(difluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromobenzal fluoride

035679-1G 035679-5G 035679-25G
101839-652EA 24.62 USD
101839-652 101839-654 102551-172
1-Bromo-2-(difluoromethyl)benzene ≥97%
1-Bromo-2-(difluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₅BrF₂
MW: 207.0154 g/mol
MDL Number: MFCD06657962
CAS Number: 845866-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR