1-(Difluoromethyl)-2,4,5-trifluorobenzene ≥97%

Supplier: Matrix Scientific

035677-1G 035677-5G
101839-644EA 106.15 USD
101839-644 101839-646
1-(Difluoromethyl)-2,4,5-trifluorobenzene ≥97%
1-(Difluoromethyl)-2,4,5-trifluorobenzene

Hazard: Irritant

Formula: C₇H₃F₅
MDL Number: MFCD08276247
CAS Number: 886510-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR