3-(Difluoromethyl)-1-fluorobenzene ≥97%

Supplier: Matrix Scientific

035675-1G 035675-5G
101839-636EA 61.54 USD
101839-636 101839-638
3-(Difluoromethyl)-1-fluorobenzene ≥97%
3-(Difluoromethyl)-1-fluorobenzene

Hazard: Irritant

Formula: C₇H₅F₃
MDL Number: MFCD06657983
CAS Number: 26029-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR